Learn More
anti-DMAP1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP203982SS
 View more versions of this product
Catalog No. NBP203982SS
Add to cart
Description
Description
DMAP1 Polyclonal antibody specifically detects DMAP1 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
DMAP1
0.5 mg/mL
Western Blot 1.0-4.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml
NP_001029196
DNA methyltransferase 1 associated protein 1, DNA methyltransferase 1-associated protein 1, DNMAP1DNMT1-associated protein 1, DNMT1 associated protein 1, DNMTAP1, EAF2, FLJ11543, KIAA1425DKFZp686L09142, MEAF2, SWC4
Rabbit
IgG
0.025 mg
Apoptosis, Chromatin Research, DNA Repair, MAP Kinase Signaling
Primary
55929
Human, Mouse, Rat, Bovine, Canine, Primate
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Purified
DMAP1
A portion of amino acids 250-300 of human DMAP1 was used as the immunogen.
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Reacts with Chimpanzee, Cow, Dog, Human, Mouse and Rat. Predicted to react with Chicken and Zebrafish. The amino acid sequence used as immunogen is 100% homologous in Chimpanzee, Dog, Human, Rat, Cow, Mouse, 88% homologous in Chicken and 77% homologous in Zebrafish.