Learn More
DMAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA1886
Description
PA1-886 detects DNA methyltransferase 1 associated protein (DMAP1) from human and mouse cells. PA1-886 has been successfully used in Western blot and Immunohistochemistry (paraffin) procedures. By Western blot, this antibody detects an ∽52 kDa band which corresponds to DMAP1 from HeLa cell nuclear extracts. PA1-886 immunizing peptide corresponds to amino acid residues 435-450 from mouse DMAP1. This sequence is completely conserved in human DMAP1. PA1-886 immunizing peptide (Cat. # PEP-118) is available for use in neutralization and control experiments.
DMAP1 Polyclonal antibody specifically detects DMAP1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
DMAP1 | |
Polyclonal | |
Unconjugated | |
DMAP1 | |
1500016M21Rik; DKFZp686L09142; Dmap1; Dmtap1; DNA methyltransferase 1 associated protein 1; DNA methyltransferase 1-associated protein 1; DNMAP1; DNMT1 associated protein 1; DNMT1 associated protein-1; DNMT1-associated protein 1; DNMTAP1; EAF2; FLJ11543; KIAA1425; MAT1-mediated transcriptional repressor; MEAF2; mKIAA1425; Mmtr; RP5-891H21.2; SWC4 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
55929, 66233 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q9JI44, Q9NPF5 | |
DMAP1 | |
Synthetic peptide corresponding to residues C-D(435) T I I D V V G A P L T R N S R(450) of mouse DMAP1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.