Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

diencephalon/mesencephalon homeobox 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human DMBX1 protein.

Manufacturer:  Abnova Corporation H00127343B01

Catalog No. 89-008-519


Add to cart

Description

Description

This gene encodes a member of the bicoid sub-family of homeodomain-containing transcription factors. The encoded protein acts as a transcription factor and may play a role in brain and sensory organ development. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq

Sequence: MQHYGVNGYSLHAMNSLSAMYNLHQQAAQQAQHAPDYRPSVHALTLAERLADIILEARYGSQHRKQRRSRTAFTAQQLEALEKTFQKTHYPDVVMRERLAMCTNLPEARVQVWFKNRRAKFRKKQRSLQKEQLQKQKEAEGSHGEGKAEAPTPDTQLDTEQPPRLPGSDPPAELHLSLSEQSASESAPEDQPDREEDPRAGAEDPKAEKSPGADSKGLGCKRGSPKADSPGSLTITPVAPGGGLLGPSHSYSSSPLSLFRLQEQFRQHMAATNNLVHYSSFEVGGPAPAAAAAAAAVPYLGVNMAPLGSLHCQSYYQSLSAAAAAHQGVWGSPLLPAPPAGLAPASATLNSKTTSIENLRLRAKQHAASLGLDTLPN
Specifications

Specifications

diencephalon/mesencephalon homeobox 1
Unconjugated
No additive
NM_172225.1
MBX|OTX3|PAXB
Mouse
50 uL
Transcription Regulation
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human DMBX1 protein.
DMBX1
Serum
DMBX1
DMBX1 (NP_757379.1, 1 a.a. ~ 377 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
127343
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit