Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DMC1, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP133688

Catalog No. NBP133688

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

DMC1
0.9mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
DMC1
Recombinant protein encompassing a sequence within the center region of human DMC1. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
disrupted meiotic cDNA1, yeast, homolog of, DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologousrecombination (yeast), DMC1 homologue, HsLim15, LIM15yeast homolog) meiosis-specific homologousrecombination, meiotic recombination protein DMC1/LIM15 homolog, MGC150472, MGC150473
Rabbit
IgG
0.1 ml
Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Homologous Recombination, Tumor Suppressors
Primary
11144
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only