Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMC1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526337

Catalog No. PIPA526337

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

DMC1 is essential for meiotic homologous recombination. Genetic recombination in meiosis plays an important role in generating diversity of genetic information. The product of this protein is structurally and evolutionary related to the products of the yeast RAD51 and E. coli RecA genes.
Specifications

Specifications

DMC1
Polyclonal
PBS with 0.09% sodium azide
Q14565
DMC1H, HsLim15, LIM15, dJ199H16.1, DMC1 homologue, disrupted meiotic cDNA1, yeast, homolog of, meiotic recombination protein DMC1/LIM15 homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11144
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
DMC1
Liquid
DMC1
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human DMC1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit