Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

doublesex and mab-3 related transcription factor 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human DMRT1 protein.

Manufacturer:  Abnova Corporation H00001761B01

Catalog No. 89-005-253


Add to cart

Description

Description

This gene is found in a cluster with two other members of the gene family, having in common a zinc finger-like DNA-binding motif (DM domain). The DM domain is an ancient, conserved component of the vertebrate sex-determining pathway that is also a key regulator of male development in flies and nematodes. This gene exhibits a gonad-specific and sexually dimorphic expression pattern. Defective testicular development and XY feminization occur when this gene is hemizygous. [provided by RefSeq

Sequence: MPNDEAFSKPSTPSEAPHAPGVPPQGRAGGFGKASGALVGAASGSSAGGSSRGGGSGSGASDLGAGSKKSPRLPKCARCRNHGYASPLKGHKRFCMWRDCQCKKCNLIAERQRVMAAQVALRRQQAQEEELGISHPIPLPSAAELLVKRENNGSNPCLMTECSGTSQPPPASVPTTAASEGRMVIQDIPAVTSRGHVENTPDLVSDSTYYSSFYQPSLFPYYNNLYNCPQYSMALAADSASGEVGNPLGGSPVKNSLRGLPGPYVPGQTGNQWQMKNMENRHAMSSQYRMHSYYPPPSYLGQSVPQFFTFEDAPSYPEARASVFSPPSSQDSGLVSLSSSSPISNKSTKAVLECEPASEPSSFTVTPVIEEDE
Specifications

Specifications

doublesex and mab-3 related transcription factor 1
Unconjugated
No additive
NM_021951.2
DMT1
Mouse
50 uL
Transcription Regulation
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human DMRT1 protein.
DMRT1
Serum
DMRT1
DMRT1 (NP_068770.2, 1 a.a. ~ 373 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1761
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit