Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMRT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA528331
Description
Recommended positive controls: K562. Predicted reactivity: Mouse (83%), Rat (81%), Pig (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DMRT1 Polyclonal antibody specifically detects DMRT1 in Human samples. It is validated for Western Blot.Specifications
DMRT1 | |
Polyclonal | |
Unconjugated | |
DMRT1 | |
CT154; DM domain expressed in testis 1; DM domain expressed in testis protein 1; Dmrt1; DMT1; Doublesex and mab-3 related transcription factor 1; doublesex- and mab-3-related transcription factor 1; Unknown (protein for MGC:134119) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1761 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.03 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y5R6 | |
DMRT1 | |
Recombinant protein fragment corresponding to a region within amino acids 78 and 339 of Human DMRT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction