Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMRT3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524027

Catalog No. PIPA524027

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May regulate transcription during sexual development.
Specifications

Specifications

DMRT3
Polyclonal
PBS with 0.09% sodium azide
Q9NQL9, Q80WT2
DMRTA3, DMRT-like family A3, doublesex- and mab-3-related transcription factor 3, testis-specific protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
240590, 58524
Western Blot
Unconjugated
DMRT3
Liquid
DMRT3
KLH conjugated synthetic peptide between 431-459 amino acids from the C-terminal region of human DMRT3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit