Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMRTA1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524343

Catalog No. PIPA524343

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

May be involved in sexual development.
Specifications

Specifications

DMRTA1
Polyclonal
PBS with 0.09% sodium azide
Q5VZB9, Q8CFG4
DMO, doublesex- and mab-3-related transcription factor A1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
242523, 63951
Western Blot
Unconjugated
DMRTA1
Liquid
DMRTA1
KLH conjugated synthetic peptide between 322-350 amino acids from the C-terminal region of human DMRTA1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit