Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Mouse anti-All, Clone: F7-26, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB3299
Description
Anti-DNA Antibody, single stranded specifimmunocytochemistry, clone F7-26 detects level of DNA and has been published and validated for use in flow cytometry, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin).
Specifications
DNA | |
Monoclonal | |
Unconjugated | |
Mouse | |
Ammonium sulfate precipitation | |
Primary | |
−80°C in undiluted aliquots for up to 12 months. | |
IgM |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
F7-26 | |
Purified Immunoglobulin. Liquid in PBS supplemented with 5% fetal bovine Serum (FBS), containing no preservatives. | |
F7-26 was generated by the immunization of mice with calf thymus single-stranded DNA. | |
50 μg | |
All species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction