Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Ligase I Mouse anti-Human, Clone: 1A9, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA126060
Description
MA1-26060 detects DNA Ligase I from human samples. MA1-26060 has been successfully used in Western blot procedures. The MA1-26060 immunogen is purified, full length, native bovine protein.
There are four distinct ligase proteins found in mammalian cells. The activity of these DNA ligases is required for the joining of DNA strand breaks that occur during DNA replication, repair and recombination. DNA ligase I migrates as a 125kDa phosphoprotein and is implicated in joining Okazaki fragments during DNA synthesis. It also has a role in the base and nucleotide excision repair pathways.Specifications
DNA Ligase I | |
Monoclonal | |
1.1 mg/mL | |
LIG1 | |
MGC117397, MGC130025 | |
Mouse | |
IgG fraction | |
RUO | |
3978 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1A9 | |
Unconjugated | |
P18858 | |
LIG1 | |
Purified, full length, native bovine protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title