Learn More
DNA Ligase I, Polyclonal, Invitrogen™
Mouse Polyclonal Antibody
Supplier:  Thermo Scientific PA123187
Catalog No. PIPA123187
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
There are four distinct ligase proteins found in mammalian cells. The activity of these DNA ligases is required for the joining of DNA strand breaks that occur during DNA replication, repair and recombination. DNA Ligase I migrates as a 125kDa phosphoprotein and is implicated in joining Okazaki fragments during DNA synthesis. It also has a role in the base and nucleotide excision repair pathways.
PA1-23187 detects DNA ligase I in Human samples.PA1-23187 has been successfully used in Western Blot procedures.Positive Control: HeLaPA1-23187 immunogen corresponds to Full-length recombinant human DNA Ligase I protein.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNA Ligase I
Polyclonal
LIG1
MGC117397, MGC130025
Mouse
50 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
P18858
LIG1
Full-length recombinant human DNA Ligase I protein
RUO
3978
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Serum
Product Suggestions
Product Suggestions