Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA-PKcs Mouse anti-Human, Unlabeled, Clone: 6, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610804

Catalog No. BDB610804


Add to cart

Description

Description

DNA-PK is a trimeric enzyme that is composed of a catalytic subunit of ~ 465kDa (DNA-PKcs) and a heterodimeric regulatory subunit of 70kDa and 86kDa. DNA-PKcs has been reported to be inactive alone and depends on the regulatory subunit f

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

DNA-PKcs
6
Unconjugated
Affinity Purified
Human DNA-PKcs aa. 874-1024
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit