Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Primase Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA129133

Catalog No. PIPA129133

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-29134 detects DP1 from rat, human, mouse samples. PA1-29134 has been successfully used in immunoprecipitation, Western blot applications. The PA1-29134 immunogen is: Synthetic peptide corresponding to the N-terminus of human DP-1 protein

The replication of DNA in eukaryotic cells is carried out by a complex chromosomal replication apparatus, in which DNA polymerase alpha and primase are two key enzymatic components. Primase, which is a heterodimer of a small subunit and a large subunit, synthesizes small RNA primers for the Okazaki fragments made during discontinuous DNA replication. The protein encoded by this gene is the small, 49 kDa primase subunit.
Specifications

Specifications

DNA Primase
Polyclonal
Unconjugated
PRIM1
Liquid
PRIM1
Purified MOUSE recombit p49 subunit of DNA primase
Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
PBS with 0.2% BSA and 0.1% sodium azide; pH 7.4
P49642, P20664
MGC12308, p49
Rabbit
IgG
500 μL
4° C, do not freeze
Primary
19075, 5557
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit