Learn More
MilliporeSigma™ anti-DNA, single stranded (ssDNA) ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB3868/DEL
Description
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Specifications
DNA, single stranded (ssDNA) | |
Unconjugated | |
Human Raji cell nuclei | |
RUO | |
Primary | |
IgG2a |
Monoclonal | |
Mouse | |
100 μg | |
Epitope Tags | |
Semi-purified |