Learn More
HDJ2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525744
Catalog No. PIPA525744
Add to cart
Description
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
It is a novel human homolog of E. coli ft.s DNAJ protein from heat shock protein 40 family. Hsp40 is involved in chaperonic protein folding HDJ-2 is cytoplasmic and nuclear membrane associated protein; upon heat shock it migrates and becomes associated with the Golgi complex, nucleolus and nuclear membrane HDJ-2 has a prenylated C-terminus and is not inducible upon heat shock.
Specifications
Specifications
HDJ2
Polyclonal
Unconjugated
DNAJA1
Liquid
DNAJA1
KLH conjugated synthetic peptide between 294-323 amino acids from the central region of human DNAJA1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P31689
DJ-2, DjA1, HDJ2, HSDJ, HSJ2, HSPF4, NEDD7, hDJ-2, HSJ-2, dnaJ homolog subfamily A member 1, dnaJ protein homolog 2, heat shock 40 kDa protein 4, heat shock protein J2, heat shock protein, DNAJ-like 2, human DnaJ protein 2, neural precursor cell expressed, developmentally down-regulated 7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3301