Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJB13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524368

Catalog No. PIPA524368

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

DNAJA13 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins.
Specifications

Specifications

DNAJB13
Polyclonal
PBS with 0.09% sodium azide
P59910
RSPH16A, TSARG5, TSARG6, 1700014P03Rik, DnaJ (Hsp40) related, subfamily B, member 13, DnaJ-like protein, dnaJ homolog subfamily B member 13, radial spoke 16 homolog A, testis and spermatogenesis cell-related protein 6, testis spermatocyte apoptosis-related gene 6 protein, testis spermat
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
374407
Immunohistochemistry, Western Blot
Unconjugated
DNAJB13
Liquid
DNAJB13
KLH conjugated synthetic peptide between 176-205 amino acids from the central region of human DNAJB13
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit