Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAJB5 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216192
Description
DNAJB5 Polyclonal antibody specifically detects DNAJB5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
DNAJB5 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
DnaJ (Hsp40) homolog, subfamily B, member 5, heat shock cognate 40, Heat shock protein cognate 40, Heat shock protein Hsp40-2, Heat shock protein Hsp40-3, HSC40, Hsc40dnaJ homolog subfamily B member 5, KIAA1045 | |
Rabbit | |
39 kDa | |
0.1 mL | |
Primary | |
Human, Mouse. Expected to cross react based on sequence identity: Rat (100%), Bovine (99%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
O75953 | |
DNAJB5 | |
Recombinant protein encompassing a sequence within the center region of human DNAJB5. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
25822 | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only