Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC11 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523982

Catalog No. PIPA523982

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Belongs to the DNAJC11 family. Three named isoforms produced by alternative splicing.
Specifications

Specifications

DNAJC11
Polyclonal
PBS with 0.09% sodium azide
Q9NVH1, Q5U458
dJ126A5.1, dnaJ homolog subfamily C member 11, novel DnaJ domain-containing protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
230935, 55735
Immunohistochemistry, Western Blot
Unconjugated
DNAJC11
Liquid
DNAJC11
KLH conjugated synthetic peptide between 27-56 amino acids from the N-terminal region of human DNAJC11
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit