Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DnaJ (Hsp40) homolog, subfamily C, member 19, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant DNAJC19.

Manufacturer:  Abnova Corporation H00131118A01

Catalog No. 89-004-785


Add to cart

Description

Description

Sequence: RYVLQAMKHMEPQVKQVFQSLPKSAFSGGYYRGGFEPKMTKREAALILGVSPTANKGKIRDAHRRIMLLNHPDKGGSPYIAAKINEAKDLLEGQAKK
Specifications

Specifications

DnaJ (Hsp40) homolog, subfamily C, member 19
Unconjugated
50% glycerol
NM_145261
TIM14|TIMM14
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
131118
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant DNAJC19.
DNAJC19
Serum
DNAJC19
DNAJC19 (NP_660304, 20 a.a. ~ 116 a.a) partial recombinant protein with GST tag.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit