Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAL4, Polyclonal, Invitrogen™

Catalog No. PIPA127771
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127771 50 μg
1 options

Catalog No. PIPA127771

Supplier: Thermo Scientific PA127771

Chicken Polyclonal Antibody

Force generating protein of respiratory cilia. Produces force towards the minus ends of microtubules. Dynein has ATPase activity (By similarity).

PA1-27771 detects DNAL4 from human, mouse, and rat samples.PA1-27771 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27499 immunogen is a synthetic peptide corresponding to human Dynein light chain 4. axonemal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAL4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene DNAL4
Gene Accession No. O96015, Q9DCM4
Gene Alias PIG27
Gene Symbols DNAL4
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Dynein light chain 4. axonemal.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10126
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.