Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNase II Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112504

Catalog No. 01-674-164


Add to cart

Description

Description

A suggested positive control for this product is human spleen lysate.

Apoptosis is characterized by several morphological nuclear changes including chromatin condensation and nuclear fragmentation. These changes are triggered by the activation of members of caspase family, caspase activated DNase, and several novel proteins including AIF and Acinus. DNase II causes both chromatin condensation and DNA fragmentation. The genes encoding human, porcine, and murine DNase II have been cloned. The DNase II gene encodes a 40 kDa proenzyme. The mature enzyme consists of two non-identical subunits, the 32 kDa (a) and 12 kDa (b) chains, generated by proteolytic processing. Overexpression of DNase II induces chromatin condensation (3). DNase II is ubiquitously expressed in human tissues.
Specifications

Specifications

DNase II
Polyclonal
PBS with 0.02% sodium azide
O00115
DNASE2A, DNL, DNL2
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1777
Western Blot
Unconjugated
DNASE2
Liquid
DNASE2
Synthetic peptide corresponding to residues 347-360 of human DNase II precursor.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit