Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC8 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525488

Catalog No. PIPA525488

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with rat based on sequence homology.

Specifications

Specifications

DNAJC8
Polyclonal
PBS with 0.09% sodium azide
O75937, Q6NZB0
HSPC331, SPF31, dnaJ homolog subfamily C member 8, splicing protein spf31
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22826, 68598
Western Blot
Unconjugated
DNAJC8
Liquid
DNAJC8
KLH conjugated synthetic peptide between 210-239 amino acids from the C-terminal region of human DNJC8
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit