Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT1 (DNA Methyl Transferase-1) Mouse anti-Human, Unlabeled, Clone: 18, BD

Mouse Monoclonal Antibody

$213.00

Specifications

Antigen DNMT1 (DNA Methyl Transferase-1)
Isotype IgG2b
Immunogen Human DNMT1 aa. 476-670
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB612618
View Documents Promotion Details
BD Biosciences
612618
50μg
Each for $213.00
Add to cart
 
BDB612619
View Documents Promotion Details
BD Biosciences
612619
150μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

DNA methylation of the 5′ position of the cytosine ring within CpG dinucleotides is essential for embryonic development. This methylation may affect transcriptional repression, formation of compact chromatin structures, X chromosome inactivation, and imprinting control. Three important DNA (Cytosine-5) methyltransferases (DNMTs) include DNMT1, DNMT3A, and DNMT3B. DNMT1 contains an N-terminal regulatory domain that may target it to replication foci, and a C-terminal catalytic domain that is homologous to bacterial cystosine 5-methylases. DNMT1 knockout mice have reduced DNA methylation, defects in imprinting, and derepression of endogenous retroviruses, and are embryonic lethal. DNMT1 is responsible for DNA methylations that occur after replication, while DNMT3A and DNMT3B act to establish new methylation patterns during embryogenesis. DNMT1 and DNMT3A are expressed in most fetal tissues, while DNMT3B is expressed only in fetal liver. All of these DNMTs are found in various adult tissues and show increased expression in some tumors and cancer cell lines. Thus, DNMT methylation of CpG dinucleotides is important for regulating DNA structure during development. This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or repoerted in the literature.

Immunofluorescence, Western Blotting

Specifications

Specifications

DNMT1 (DNA Methyl Transferase-1)
Human DNMT1 aa. 476-670
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
DNA Methyl Transferase-1
IgG2b
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
18
250μg/mL
Store undiluted at -20°C.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit