Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DNMT3A, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $293.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 10-50 of human Dnmt3a was used as the immunogen.
Antigen DNMT3A
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224471SS
View Documents
Novus Biologicals
NBP224471SS
0.025 mg
Each for $119.60
Add to cart
 
NBP224471
View Documents
Novus Biologicals
NBP224471
0.1 mg
Each for $293.00
Add to cart
 
Description

Description

DNMT3A Polyclonal antibody specifically detects DNMT3A in Human, Primate samples. It is validated for Western Blot.
Specifications

Specifications

Rabbit
A portion of amino acids 10-50 of human Dnmt3a was used as the immunogen.
Polyclonal
Primary
Western Blot
Q9Y6K1
PBS, 0.05% BSA with 0.05% Sodium Azide
IgG
DNMT3A
Affinity Purified
RUO
This STAT6 antibody will cross-react with mouse and rat, based on NCBI BLAST.
1788
0.5 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit