Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511159

Catalog No. PIPA511159

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken, mouse and rat based on sequence homology.

CpG methylation is an epigenetic modification that is important for embryonic development, imprinting, and X-chromosome inactivation. Studies in mice have demonstrated that DNA methylation is required for mammalian development. Dnmt3a is a DNA methyltransferase that is thought to function in de novo methylation, rather than maintece methylation. The protein localizes to the cytoplasm and nucleus and its expression is developmentally regulated.
Specifications

Specifications

Dnmt3a
Polyclonal
PBS with 0.09% sodium azide
Q9Y6K1, O88508
DNA (cytosine-5)-methyltransferase 3A; Dnmt3a, DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, M.HsaIIIA
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
13435, 1788
Immunohistochemistry, Western Blot
Unconjugated
DNMT3A
Liquid
DNMT3A
KLH conjugated synthetic peptide between 457-486 amino acids from the central region of human Dnmt3a
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit