Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-DOG IGG (GAMMA) AB HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200289
Affinity purified polyclonal secondary antibody against Dog IgG, heavy chains (gamma) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form.
Catalog No. 50-669-44