Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-DOG IGG (H+L) ANTIBODY  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52100195
Affinity purified polyclonal antibody to Dog IgG, both heavy and light chains (whole IgG) made in Goat. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-668-33