Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOK5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514756

Catalog No. PIPA514756

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

DOK5 is a member of the DOK family of membrane proteins, which are adapter proteins involved in signal transduction. The encoded protein interacts with phosphorylated receptor tyrosine kinases to mediate neurite outgrowth and activation of the MAP kinase pathway. In contrast to other DOK family proteins, this protein does not interact with RASGAP.
Specifications

Specifications

DOK5
Polyclonal
PBS with 0.09% sodium azide
Q9P104, Q91ZM9
Docking protein 5; Downstream of tyrosine kinase 5; IRS6; Protein dok-5;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55816, 76829
Immunohistochemistry, Western Blot
Unconjugated
DOK5
Liquid
DOK5
KLH conjugated synthetic peptide between 25-54 amino acids from the N-terminal region of human DOK5
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit