Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOM3Z Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513587

Catalog No. PIPA513587

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The function of DOM3Z is unknown, but its ubiquitous expression and conservation in both simple and complex eukaryotes suggests that its gene may be a housekeeping gene.
Specifications

Specifications

DOM3Z
Polyclonal
PBS with 0.09% sodium azide
O77932
Protein Dom3Z; Dom-3 homolog Z; NG6; DOM3L; DOM3Z
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1797
Flow Cytometry, Western Blot
Unconjugated
DXO
Liquid
DXO
KLH conjugated synthetic peptide between 73-99 amino acids from the N-terminal region of human DOM3Z
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit