Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Donkey IgG, Isotype Control, Novus Biologicals™

Donkey Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19704810MG

Catalog No. NBP19704810


Add to cart

Description

Description

IgG antibody specifically detects IgG in Donkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Dot Blot.
Specifications

Specifications

IgG
Unconjugated
Lyophilized
Donkey
Ion exchange chromatography
RUO
Isotype Control
This product was prepared from normal serum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Donkey IgG and anti-Donkey Serum.
Donkey
ELISA, Immunoprecipitation, Western Blot
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: None. with 0.01% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
IgG
10 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only