Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DONSON Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525568

Catalog No. PIPA525568

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

DONSON lies downstream of the SON gene and spans 10 kbon chromosome 21. The function of this gene is unknown.
Specifications

Specifications

DONSON
Polyclonal
PBS with 0.09% sodium azide
Q9NYP3
B17, C21orf60, C2TA, MHC class II transactivator type I, protein downstream neighbor of Son
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29980
Flow Cytometry, Western Blot
Unconjugated
DONSON
Liquid
DONSON
KLH conjugated synthetic peptide between 368-398 amino acids from the central region of human DONSON
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit