Learn More
DOPA Decarboxylase Rabbit anti-Bovine, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA14651
Catalog No. PA14651
Add to cart
Description
Description
This antibody is specific for the ~55 kDa DDC protein in Western blots of rat adrenal medulla.
DOPA Decarboxylase (aromatic L-amino acid decarboxylase, AADC; DDC) catalyzes the second reaction in the biosynthesis of catecholamines and serotonin (Waymire and Haycock, 2002; Berry et al., 1996; Haycock et al., 2003). It is also involved in the biosynthesis of trace amines. DDC antibodies can therefore be used as markers for dopaminergic, noradrenergic and serotonergic neurons in a variety of applications including depression, schizophrenia, Parkinson€™s disease and drug abuse (Kish et al., 2001; Zhu et al., 2000; Zhu et al., 1999).
Specifications
Specifications
DOPA Decarboxylase
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
P27718, P14173
aromatic L-amino decarboxylase, AADC, DDC
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
24311, 280762
Western Blot
Unconjugated
Ddc
Liquid
DDC
SDS denatured, recombit bovine aromatic DOPA decarboxylase expressed in E. coli and purified from inclusion bodies
Affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Rat