Learn More
Dopamine beta Hydroxylase Sheep anti-Human, Non-human primate, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA14655
Catalog No. PA14655
Add to cart
Description
Description
This antibody is specific for the ~75 kDa DBH protein in Western blots of human adrenal medulla.
DBH catalyzes the conversion of dopamine to norepinephrine and serves as a marker of noradrenergic cells. DBH antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson’s disease and drug abuse (Kish et al., 2001; Zhu et al., 2000; Zhu et al., 1999). The expression of DBH is also elevated during stress (Sabban and Kvetsky, 2001).
Specifications
Specifications
Dopamine beta Hydroxylase
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
P09172
DBH, Dopamine beta monooxygenase
Sheep
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1621
Western Blot
Unconjugated
DBH
Liquid
DBH
Synthetic peptide corresponding to the N-terminal region of human DBH conjugated to KLH
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Non-human Primate