Learn More
DP103/Gemin3 Mouse anti-Human, Unlabeled, Clone: 2, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 612153
Catalog No. BDB612153
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
DEAD box proteins, a family of putative RNA helicases, are characterized by eight conserved amino acid motifs that are arranged in a core region as found in the prototypical member of the family, eIF-4A. The family's name is derived from the amino acid sequence Asp-Glu-Ala-Asp (DEAD) that is located within the ATP hydrolysis motif. DEAD box proteins have been implicated in translation initiation and RNA splicing, degradation, and stability. The DEAD box protein, DP103, contains seven N-terminal helicase motifs characteristic of DEAD box proteins followed by an SMN interaction domain (SID). DP103 mRNA has been reported to be widely expressed, and DP103 protein is found in the nucleus and cytoplasm. SMN, the gene mutated in spinal muscular atrophy, forms a 20S nuclear complex that includes DP103, SIP1 (SMN-interacting protein 1), and snRNPs. DP103 also co-localizes with SMN and SIP1 to nuclear bodies called gems. In addition, DP103 interacts with the proximal repressor domain of steroidogenic factor-1, a nuclear receptor essential for development of the gonads, adrenal gland, and hypothalamic nuclei. Thus, DP103 may have roles in SMN complex modulation of RNA splicing and in transcriptional repression.
Western Blotting
Specifications
Specifications
DP103/Gemin3
2
Unconjugated
Murine
IgG2b
RUO
Primary
Human
Western Blot
250μg/mL
Affinity Purified
Human DP103 aa. 667-783
150μg
Cell Biology
Monoclonal