Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DP103 (Gemin3; DEAD box Protein-103; Ddx20) Mouse anti-Human, Unlabeled, Clone: 2, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612152

Catalog No. BDB612152


Add to cart

Description

Description

DEAD box proteins, a family of putative RNA helicases, are characterized by eight conserved amino acid motifs that are arranged in a core region as found in the prototypical member of the family, eIF-4A. The family's name is derived from the amino acid sequence Asp-Glu-Ala-Asp (DEAD) that is located within the ATP hydrolysis motif. DEAD box proteins have been implicated in translation initiation and RNA splicing, degradation, and stability. The DEAD box protein, DP103, contains seven N-terminal helicase motifs characteristic of DEAD box proteins followed by an SMN interaction domain (SID). DP103 mRNA has been reported to be widely expressed, and DP103 protein is found in the nucleus and cytoplasm. SMN, the gene mutated in spinal muscular atrophy, forms a 20S nuclear complex that includes DP103, SIP1 (SMN-interacting protein 1), and snRNPs. DP103 also co-localizes with SMN and SIP1 to nuclear bodies called gems. In
addition, DP103 interacts with the proximal repressor domain of steroidogenic factor-1, a nuclear receptor essential for development of the gonads, adrenal gland, and hypothalamic nuclei. Thus, DP103 may have roles in SMN complex modulation of RNA splicing and in transcriptional repression.

Western Blotting

Specifications

Specifications

DP103 (Gemin3; DEAD box Protein-103; Ddx20)
2
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Gemin3; DEAD box Protein-103; Ddx20
Affinity Purified
Human DP103 aa. 667-783
50μg
Cell Biology
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit