Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPEP3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524277

Catalog No. PIPA524277

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a membrane-bound glycoprotein from the family of dipeptidases involved in hydrolytic metabolism of various dipeptides, including penem and carbapenem beta-lactam antibiotics. This gene is located on chromosome 16 in a cluster with another member of this family. Alternatively spliced transcript variants that encode different isoforms have been found for this gene.
Specifications

Specifications

DPEP3
Polyclonal
PBS with 0.09% sodium azide
Q9H4B8, Q9DA79
MBD3, membrane-bound dipeptidase 3, metallopeptidase (family M19)
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64180, 71854
Western Blot
Unconjugated
DPEP3
Liquid
DPEP3
KLH conjugated synthetic peptide between 391-420 amino acids from the C-terminal region of human DPEP3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit