Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPF3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525345

Catalog No. PIPA525345

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken based on sequence homology.

Specifications

Specifications

DPF3
Polyclonal
PBS with 0.09% sodium azide
Q92784
CERD4, 2810403B03Rik, BAF45C, BRG1-associated factor 45C, cer-d4 homolog, zinc finger protein DPF3, zinc finger protein cer-d4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8110
Western Blot
Unconjugated
DPF3
Liquid
DPF3
KLH conjugated synthetic peptide between 143-171 amino acids from the central region of human DPF3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit