Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPH2 homolog (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human DPH2 protein.

Manufacturer:  Abnova Corporation H00001802B01

Catalog No. 89-005-258


Add to cart

Description

Description

This gene is one of two human genes similar to the yeast gene dph2. The yeast gene was identified by its ability to complement a diphthamide mutant strain, and thus probably functions in diphthamide biosynthesis. Diphthamide is a post-translationally modified histidine residue present in elongation factor 2 (EF2) that is the target of diphtheria toxin ADP-ribosylation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MESMFSSPAEAALQRETGVPGLLTPLPDLDGVYELERVAGFVRDLGCERVALQFPDQLLGDAVAVAARLEETTGSKMFILGDTAYGSCCVDVLGAEQAGAQALIHFGPACLSPPARPLPVAFVLRQRSVALELCVKAFEAQNPDPKAPVVLLSEPACAHALEALATLLRPRYLDLLVSSPAFPQPVGSLSPEPMPLERFGRRFPLAPGRRLEEYGAFYVGGSKASPDPDLDPDLSRLLLGWAPGQPFSSCCPDTGKTQDEGARAGRLRARRRYLVERARDARVVGLLAGTLGVAQHREALAHLRNLTQAAGKRSYVLALGRPTPAKLANFPEVDVFVLLACPLGALAPQLSGSFFQPILAPCELEAACNPAWPPPGLAPHLTHYADLLPGSPFHVALPPPESELWETPDVSLITGDLRPPPAWKSSNDHGSLALTPRPQLELAESSPAASFLSSRSWQGLEPRLGQTPVTEAVSGRRGIAIAYEDEGSG
Specifications

Specifications

DPH2 homolog (S. cerevisiae)
Unconjugated
No additive
NM_001384.4
DPH2L2
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1802
Western Blot
Mouse polyclonal antibody raised against a full-length human DPH2 protein.
DPH2
Serum
DPH2
DPH2 (NP_001375.2, 1 a.a. ~ 489 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit