Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DPM2, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128470

Catalog No. NBP128470


Add to cart

Description

Description

DPM2 Polyclonal antibody specifically detects DPM2 in Mouse samples. It is validated for Peptide ELISA.
Specifications

Specifications

DPM2
0.5 mg/mL
Peptide ELISA Detection limit 1:16000
NP_034203.1
dolichol phosphate-mannose biosynthesis regulatory protein, dolichol phosphate-mannose synthase 2, dolichyl-phosphate mannosyltransferase polypeptide 2, regulatory subunit, FLJ80013, MGC111193, MGC21559
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
DPM2
Peptide with sequence C-YVMLKSQKITKKAQ corresponding to C-Terminus according to NP_034203.1.
Immunogen affinity purified
RUO
Primary
8818
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only