Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPP10 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14385

Catalog No. PA14385


Add to cart

Description

Description

PA1-4385 detects DPP10 in human samples. PA1-4385 has been successfully used in Western blot procedures. By Western blot, this antibody detects a ~75 kDa protein representing DPP10. Oligermization and differential glycosylation produces other bands as observed with other DPP family members. PA1-4385 immunizing peptide corresponds to amino acid residues 521-539 of human DPP10.

A novel gene, DPP10, encodes a homolog of dipeptidyl peptidases (DPPs) that cleave terminal dipeptides from cytokines and chemokines, and it presents a potential new target for asthma therapy. The DPP10 protein shares features with members of the S9B family of DPP serine proteases, which includes DPP4, a widely expressed enzyme that plays a central role in chemokine processing as part of the innate immune system. The locus displays a complex pattern of transcript splicing, with eight alternate first axons; four of which localize within the LD block strongly associated with asthma.
Specifications

Specifications

DPP10
Polyclonal
Unconjugated
DPP10
Liquid
DPP10
Synthetic peptide: L(521) K E A I L K K K I G K P E I K I L H(539)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8N608
Inactive dipeptidyl peptidase 10, Dipeptidyl peptidase X, Dipeptidyl peptidase-like protein 2, DPL2, Dipeptidyl peptidase IV-related protein 3, DPRP-3
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
57628
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit