Learn More
DPP8 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA513431
Catalog No. PIPA513431
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This antibody is predicted to react with mouse based on sequence homology.
DPP8 is a member of the peptidase S9B family, a small family of dipeptidyl peptidases that are able to cleave peptide substrates at a prolyl bond. The protein shares similarity with dipeptidyl peptidase IV in that it is ubiquitously expressed, and hydrolyzes the same substrates. These similarities suggest that, like dipeptidyl peptidase IV, this protein may play a role in T-cell activation and immune function.
Specifications
Specifications
DPP8
Polyclonal
Unconjugated
DPP8
Liquid
DPP8
KLH conjugated synthetic peptide between 846-876 amino acids from the C-terminal region of human DPP8
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
2 mg/mL
PBS with 0.09% sodium azide
Q6V1X1
Dipeptidyl peptidase 8; Dipeptidyl peptidase VIII; DP8; Prolyl dipeptidase DPP8; Dipeptidyl peptidase IV-related protein 1; DPRP-1; MSTP141; DPRP1; DPP8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54878