Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPP8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525893

Catalog No. PIPA525893

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

DPP8 is a member of the peptidase S9B family, a small family of dipeptidyl peptidases that are able to cleave peptide substrates at a prolyl bond. The protein shares similarity with dipeptidyl peptidase IV in that it is ubiquitously expressed, and hydrolyzes the same substrates. These similarities suggest that, like dipeptidyl peptidase IV, this protein may play a role in T-cell activation and immune function.
Specifications

Specifications

DPP8
Polyclonal
PBS with 0.09% sodium azide
Q6V1X1
DP8, DPRP1, MSTP141, DPP VIII, DPRP-1, dipeptidyl peptidase 8, dipeptidyl peptidase IV-related protein 1, dipeptidyl peptidase IV-related protein-1, dipeptidyl peptidase VIII, dipeptidylpeptidase 8, prolyl dipeptidase DPP8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54878
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
DPP8
Liquid
DPP8
KLH conjugated synthetic peptide between 23-52 amino acids from the N-terminal region of human DPP8
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit