Learn More
DPP9, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127587
Description
This gene encodes a protein that is a member of the S9B family in clan SC of the serine proteases. The protein has been shown to have post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Although the activity of this protein is similar to that of dipeptidyl peptidase 4 (DPP4), it does not appear to be membrane bound. In general, dipeptidyl peptidases appear to be involved in the regulation of the activity of their substrates and have been linked to a variety of diseases including type 2 diabetes, obesity and cancer. Several transcript variants of this gene have been described but not fully characterized.
PA1-27587 detects DPRP2 from human samples.PA1-27587 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human R26984_1.Specifications
DPRP2 | |
Polyclonal | |
DPP9 | |
DKFZp762F117, DPRP2, FLJ16073 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
91039 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Western Blot | |
Unconjugated | |
Q86TI2 | |
DPP9 | |
Synthetic peptide corresponding to human R26984_1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgY |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.