Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPY19L1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524345

Catalog No. PIPA524345

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Belongs to the DPY19 family. There are two named isoforms. The specific function of DPY19L1 is not yet known.
Specifications

Specifications

DPY19L1
Polyclonal
PBS with 0.09% sodium azide
Q2PZI1
DPY-19-like protein 1, protein dpy-19 homolog 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23333
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
DPY19L1
Liquid
DPY19L1
KLH conjugated synthetic peptide between 557-586 amino acids from the C-terminal region of human DPY19L1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit