Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DPY30, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232201

Catalog No. NBP232201


Add to cart

Description

Description

DPY30 Polyclonal antibody specifically detects DPY30 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifications

Specifications

DPY30
1.0 mg/mL
Immunoprecipitation 2-10 mg/mg, Immunohistochemistry-Paraffin 1:1000-1:5000, Western Blot (Negative)
NP_115963.1
dpy-30 homolog (C. elegans), dpy-30L, Dpy-30-like protein, HDPY-30, protein dpy-30 homolog, Saf19
Rabbit
IgG
100 ul
modENCODE Antibodies
Primary
84661
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
DPY30
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human DPY-30 Homolog using the numbering given in entry NP_115963.1 (GeneID 84661).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only