Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPYSL5, Mouse, Clone: 2G4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DPYSL5.
Supplier: Abnova Corporation H00056896M02
Description
Members of the CRMP family, such as DPYSL5, are believed to play a role in growth cone guidance during neural development.[supplied by OMIM
Sequence: SFPDTVYKKLVQREKTLKVRGVDRTPYLGDVAVVVHPGKKEMGTPLADTPTRPVTRHGGMRDLHESSFSLSGSQIDDHVPKRASARILAPPGGRSSGIWSpecifications
DPYSL5 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_020134 | |
DPYSL5 | |
DPYSL5 (NP_064519, 466 a.a. ∼ 564 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Western Blot | |
2G4 | |
Mouse monoclonal antibody raised against a partial recombinant DPYSL5. | |
DPYSL5 | |
CRAM/CRMP-5/CRMP5/FLJ45383/Ulip6 | |
Mouse | |
Affinity chromatography | |
RUO | |
56896 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction