Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130533

Catalog No. PA130533


Add to cart

Description

Description

PA1-30533 detects DR3 from human samples. PA1-30533 has been successfully used in Western blot applications. The PA1-30533 immunogen is: Peptide from extracellular domain of human DR3 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

DR3 (also known as Apo3, WSL-1, TRAMP, and LARD) is expressed on lymphocytes. DR3 is one of death domain containing receptors and shows close sequence similarity to TNF receptor I. Upon overexpression, DR3 triggers responses that resemble those of TNFRI,
Specifications

Specifications

DR3
Polyclonal
Unconjugated
TNFRSF25
Liquid
TNFRSF25
Peptide from extracellular domain of human DR3
Ion-exchange chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q93038
RP4-650H14.2, APO-3, DDR3, DR3, LARD, TNFRSF12, TR3, TRAMP, WSL-1, WSL-LR
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
8718
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit