Learn More
MilliporeSigma™ anti-DR4/TRAIL-R1; Polyclonal; ™ (Upstate™)
Supplier: MilliporeSigma™ 06744/DEL
Description
Western Blotting
Specifications
TRAIL-R1/DR4 | |
Unconjugated | |
peptide (C-RPAREASPRLRVHK) corresponding to amino acids 77-90 of human DR4/TRAIL-R1 | |
RUO | |
Primary | |
Semi-purified |
Polyclonal | |
Rabbit | |
200 μg | |
Apoptosis | |
Human, Mouse, Rat | |