Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR6 Mouse anti-Human, PE, Clone: DR-6-04-EC, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA119720

Catalog No. 01-676-742


Add to cart

Description

Description

CD358 / DR6 (Death Receptor-6) is a type I transmembrane protein of the TNF receptor superfamily, expressed in most human tissues and able of inducing apoptosis through its cytoplasmic death domain. Unlike TNFR1 and Fas, DR6 induces apoptosis independently of FADD adaptor. In immune system, DR6 serves as an important regulator for CD4+ T cell proliferation and Th differentiation, and provides also a regulatory mechanism for B cell activation and humoral immune responses.

Specifications

Specifications

DR6
Monoclonal
0.1 mg/mL
Assay Dependent
TNFRSF21
Liquid
TNFRSF21
Fusion protein representing amino acids 42-335 (extracellular part) of human DR6 linked to the Fc portion of human IgG1.
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
DR-6-04-EC
PE
PBS with 0.2% BSA and 15mM sodium azide
O75509
Death receptor-6
Mouse
IgG1
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
27242
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit